Anti-ARPC1B Rabbit Polyclonal Antibody
Catalog # ABGEAP8754C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Actin related protein 2/3 complex, subunit 1B, 41kDa
- Antigen symbol:ARPC1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Center
- Gene ID:6217
- Antigen synonyms:p40-ARC|ARPC1B|ARC41|ARP2/3 protein complex subunit p41|actin-related protein 2/3 complex subunit 1B|p41-ARC|arp2/3 complex 41 kDa subunit
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:KERFAGVDIRVRVKGC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This RPS16 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 45-74 amino acids from the Central region of human RPS16.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ARPC1B (actin related protein 2/3 complex, subunit 1B, 41kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human