Anti-CA2 Rabbit Polyclonal Antibody
ABGEAP7307A
: Abgent
- Antibody type:Primary
- Antigen name:Carbonic Anhydrase 2
- Antigen symbol:CA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:CAC|Carbonic dehydratase|CA-II|Carbonate dehydratase II|HEL-76|Carbonic anhydrase II|Carbonic anhydrase C|Car2|Carbonic anhydrase B|CAII
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CA2 (carbonic anhydrase II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse