Anti-PLK2 Rabbit Polyclonal Antibody
Catalog # ABGEAP7252A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Polo-like kinase 2
- Antigen symbol:PLK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal (375–425 aa)
- Cross adsorption:No
- Antigen synonyms:PLK-2|Serine/threonine-protein kinase PLK2|Serum-inducible kinase|PLK2|Serine/threonine-protein kinase SNK|Polo-like kinase 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PLK2 (polo-like kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat