Order Entry
Italy
ContactUsLinkComponent
 
Anti-EDNRB Rabbit Polyclonal Antibody
Catalog # ABGEAP6789B
Supplier:  Abgent
undefined
Anti-EDNRB Rabbit Polyclonal Antibody
Catalog # ABGEAP6789B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Endothelin receptor type B
  • Antigen symbol:
    EDNRB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Antigen synonyms:
    ETB1|ETBR|ET-BR|WS4A|ABCDS|HSCR|ET-B|HSCR2|ETB|ETRB
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: EDNRB (endothelin receptor type B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse