Order Entry
Italy
ContactUsLinkComponent
Anti-MAGED1 Rabbit Polyclonal Antibody
Anti-MAGED1 Rabbit Polyclonal Antibody
Catalog # ABGEAP6176A
Supplier:  Abgent
undefined
Anti-MAGED1 Rabbit Polyclonal Antibody
Catalog # ABGEAP6176A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Melanoma Antigen Family D1
  • Antigen symbol:
    MAGED1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    Center (466–481 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    ARF GTPase-activating protein GIT1|Cat-1|GIT20|G protein-coupled receptor kinase-interactor 1|GRK-interacting protein 1|Cool-associated and tyrosine-phosphorylated protein 1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MAGED1 (melanoma antigen family D, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human