Anti-FDFT1 Rabbit Polyclonal Antibody
Catalog # ABGEAP2417B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Farnesyl-diphosphate farnesyltransferase 1
- Antigen symbol:FDFT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Center ( 180–195 aa)
- Cross adsorption:No
- Antigen synonyms:Farnesyl-diphosphate farnesyltransferase|Squalene synthetase|FPP:FPP farnesyltransferase|SS|SQS
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: FDFT1 (farnesyl-diphosphate farnesyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human