Order Entry
Italy
ContactUsLinkComponent
 
Anti-SLIT1 Rabbit Polyclonal Antibody
  ABGEAP13374A
 :  Abgent
undefined
Anti-SLIT1 Rabbit Polyclonal Antibody
  ABGEAP13374A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Slit Guidance Ligand 1
  • Antigen symbol:
    SLIT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-Terminal
  • Gene ID:
    6585
  • Antigen synonyms:
    Slit homolog 1 protein|SLIT 3|SLIL1|MEGF4|MEGF 4|Slit homolog 1|Slit-1|SLIT1_HUMAN.|Multiple epidermal growth factor like domains protein 4|KIAA0813|Multiple EGF like domains protein 4|Slit1|Slit 1|SLIT3|SLIL 1|Multiple epidermal growth factor like domains 4
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molecular weight:
    167.926 kDa
  • Sequence:
    AVERGAFDDMKELERC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This SLIT1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 84-113 amino acids from the N-terminal region of human SLIT1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: SLIT1 (slit homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse