Anti-SLC36A1 Rabbit Polyclonal Antibody
ABGEAP12170A
: Abgent
- Antibody type:Primary
- Antigen name:Solute Carrier Family 36 Member A1
- Antigen symbol:Solute carrier family 36, member 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:Trypsin 1|TRYP1|TRY1|TRP1|TRY4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: SLC36A1 (solute carrier family 36 (proton/amino acid symporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human