Order Entry
Italy
ContactUsLinkComponent
Anti-SLC25A12 Rabbit Polyclonal Antibody
Anti-SLC25A12 Rabbit Polyclonal Antibody
Catalog # ABGEAP10273A
Supplier:  Abgent
Anti-SLC25A12 Rabbit Polyclonal Antibody
Catalog # ABGEAP10273A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 25 Member A12
  • Antigen symbol:
    Solute carrier family 25, member 12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-Terminal ( 1–50 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    solute carrier family 25|ARALAR1|CMC1_HUMAN|Araceli hiperlarga|Mitochondrial aspartate glutamate carrier 1|Calcium-binding mitochondrial carrier protein Aralar1|Solute carrier family 25 member 12|member 12.|ARALAR|Calcium binding mitochondrial carrier superfamily member|AGC1|SLC25A12
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SLC25A12 (solute carrier family 25 (aspartate/glutamate carrier), member 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse