Order Entry
Italy
ContactUsLinkComponent
Anti-CHEK1 Mouse Monoclonal Antibody
Anti-CHEK1 Mouse Monoclonal Antibody
Catalog # ABGEAM7401A
Supplier:  Abgent
Anti-CHEK1 Mouse Monoclonal Antibody
Catalog # ABGEAM7401A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Checkpoint Kinase 1
  • Antigen symbol:
    CHEK1
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-Terminal ( 1–50 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    kinase Chk1|CHK1|EC 2.7.11.1|CHEK1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse