Anti-VN1R1 Rabbit Polyclonal Antibody
AVIVARP65524_P050
- Antibody type:Primary
- Antigen name:Vomeronasal 1 receptor 1
- Antigen symbol:VN1R1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:57191
- Antigen synonyms:V1RL1|ZVNH1|ZVNR1|VNR19I1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: VN1R1 (vomeronasal 1 receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human