Anti-TFPT Rabbit Polyclonal Antibody
Catalog # AVIVARP65109_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:TCF3 (E2A) fusion partner (in childhood Leukemia)
- Antigen symbol:TFPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:29844
- Antigen synonyms:amida|INO80F|FB1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TFPT (TCF3 (E2A) fusion partner (in childhood Leukemia))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human