Order Entry
Italy
ContactUsLinkComponent
Anti-OSCAR Rabbit Polyclonal Antibody
Anti-OSCAR Rabbit Polyclonal Antibody
Catalog # AVIVARP63083_P050
undefined
Anti-OSCAR Rabbit Polyclonal Antibody
Catalog # AVIVARP63083_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    osteoclast associated, immunoglobulin-like receptor
  • Antigen symbol:
    OSCAR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    126014
  • Antigen synonyms:
    PIgR-3|PIGR3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: OSCAR (osteoclast associated, immunoglobulin-like receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human