Anti-ALKBH1 Rabbit Polyclonal Antibody
AVIVARP61830_P050
- Antibody type:Primary
- Antigen name:alkB homolog 1, histone H2A dioxygenase
- Antigen symbol:ALKBH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:8846
- Antigen synonyms:ABH|hABH|alkB|ABH1|ALKBH
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ALKBH1 (alkB, alkylation repair homolog 1 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: