Anti-RAB5A Rabbit Polyclonal Antibody
Catalog # AVIVARP61181_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:RAB5A, member RAS oncogene family
- Antigen symbol:RAB5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:271457
- Antigen synonyms:AI663973|AU021172|2410015H04Rik|nnyRab5a
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RAB5A (RAB5A, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: