Anti-GPI Rabbit Polyclonal Antibody
Catalog # AVIVARP61088_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Glucose-6-phosphate isomerase
- Antigen symbol:GPI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:2821
- Antigen synonyms:PGI|GNPI|SA36|AMF|PHI|NLK|SA-36
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GPI (glucose-6-phosphate isomerase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: