Order Entry
Italy
ContactUsLinkComponent
Anti-SERPINB6 Rabbit Polyclonal Antibody
Anti-SERPINB6 Rabbit Polyclonal Antibody
Catalog # AVIVARP59249_P050
undefined
Anti-SERPINB6 Rabbit Polyclonal Antibody
Catalog # AVIVARP59249_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Serpin peptidase inhibitor, clade B (ovalbumin), member 6
  • Antigen symbol:
    SERPINB6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    5269
  • Antigen synonyms:
    SPI3|DFNB91|PTI|MSTP057|PI-6|PI6|CAP
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: SERPINB6 (serpin peptidase inhibitor, clade B (ovalbumin), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: