Anti-RALGPS2 Rabbit Polyclonal Antibody
Catalog # AVIVARP57092_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Ral GEF With PH Domain And SH3 Binding Motif 2
- Antigen symbol:RALGPS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:55103
- Antigen synonyms:dJ595C2.1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RALGPS2 (Ral GEF with PH domain and SH3 binding motif 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: