Anti-LCP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP54687_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:lymphocyte cytosolic protein 1 (L-plastin)
- Antigen symbol:LCP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3936
- Antigen synonyms:HEL-S-37|LPL|L-PLASTIN|LC64P|CP64|PLS2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: LCP1 (lymphocyte cytosolic protein 1 (L-plastin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: