Anti-TNP1 Rabbit Polyclonal Antibody
AVIVARP53607_P050
- Antibody type:Primary
- Antigen name:transition protein 1 (during histone to protamine replacement)
- Antigen symbol:TNP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:7141
- Antigen synonyms:TP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TNP1 (transition protein 1 (during histone to protamine replacement))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: