Anti-PNMA2 Rabbit Polyclonal Antibody
AVIVARP52314_P050
- Antibody type:Primary
- Antigen name:paraneoplastic Ma antigen 2
- Antigen symbol:PNMA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:10687
- Antigen synonyms:RGAG2|MA2|MM2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PNMA2 (paraneoplastic Ma antigen 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: