Anti-MAP3K2 Rabbit Polyclonal Antibody
AVIVARP49223_P050
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase kinase 2
- Antigen symbol:MAP3K2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10746
- Antigen synonyms:MEKK2B|MEKK2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: MAP3K2 (mitogen-activated protein kinase kinase kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: