Order Entry
Italy
ContactUsLinkComponent
Anti-SSR2 Rabbit Polyclonal Antibody
Anti-SSR2 Rabbit Polyclonal Antibody
Catalog # AVIVARP45431_P050
Anti-SSR2 Rabbit Polyclonal Antibody
Catalog # AVIVARP45431_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    signal sequence receptor, beta
  • Antigen symbol:
    SSR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    66256
  • Antigen synonyms:
    TRAPbeta|TRAPB|AU020133|AI315033|TLAP|1500032E05Rik
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: SSR2 (signal sequence receptor, beta (translocon-associated protein beta))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: