Order Entry
Italy
ContactUsLinkComponent
Anti-MGRN1 Rabbit Polyclonal Antibody
Anti-MGRN1 Rabbit Polyclonal Antibody
Catalog # AVIVARP43186_P050
Anti-MGRN1 Rabbit Polyclonal Antibody
Catalog # AVIVARP43186_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    mahogunin ring finger 1, E3 ubiquitin protein ligase
  • Antigen symbol:
    MGRN1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    23295
  • Antigen synonyms:
    RNF156
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: MGRN1 (mahogunin ring finger 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: