Anti-GNAS Rabbit Polyclonal Antibody
AVIVARP42693_P050
- Antibody type:Primary
- Antigen name:GNAS complex locus
- Antigen symbol:GNAS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2778
- Antigen synonyms:NESP|GSP|POH|GPSA|SCG6|GSA|C20orf45|AHO|GNAS1|SgVI
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GNAS (GNAS complex locus)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: