Anti-ANXA11 Rabbit Polyclonal Antibody
AVIVARP36586_T100
- Antibody type:Primary
- Antigen name:Annexin A11
- Antigen symbol:ANXA11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:311
- Antigen synonyms:ANX11|CAP50
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: ANXA11 (annexin A11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: