Anti-SOX11 Rabbit Polyclonal Antibody
Catalog # AVIVARP33328_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:SRY (sex determining region Y)-box 11
- Antigen symbol:SOX11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:6664
- Antigen synonyms:MRD27
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SOX11 (SRY (sex determining region Y)-box 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: