Anti-HOXD1 Rabbit Polyclonal Antibody
Catalog # AVIVARP32644_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:homeobox D1
- Antigen symbol:HOXD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3231
- Antigen synonyms:Hox-4.7|HOX4G|HOX4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HOXD1 (homeobox D1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: