Anti-DRAP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP31488_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:DR1-associated protein 1 (negative cofactor 2 alpha)
- Antigen symbol:DRAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10589
- Antigen synonyms:NC2-alpha
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: DRAP1 (DR1-associated protein 1 (negative cofactor 2 alpha))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: