Anti-IL27RA Rabbit Polyclonal Antibody
Catalog # BOSSBS-2711R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 27 Receptor Alpha
- Antigen symbol:IL27RA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:WSX1|Interleukin-27 receptor subunit alpha|zcytor1|IL27R|UNQ296/PRO336|CRL1|TCCR|IL-27RA|Cytokine receptor WSX-1|IL-27 receptor subunit alpha
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: IL27RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat