Anti-EGR1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1076R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Early growth response protein 1
- Antigen symbol:EGR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:nerve growth factor-induced protein A|TIS8|AT225|ZNF225|ZIF-268|NGFI-A|G0S30|KROX-24
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: EGR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat