Anti-TGF beta 1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-0086R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Transforming Growth Factor beta 1
- Antigen symbol:TGFB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:CED|Transforming growth factor beta-1|TGFbeta|Latency-associated peptide|TGFB|DPD1|LAP|TGFB1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: TGF beta 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat