Handheld digital microscopes, Dino-Lite Medical, TrichoScope
These handheld microscopes are ideal tools for studying the scalp, hair shaft and the hair itself in detail (trichoscopy). Images made with the TrichoScope can help the specialists to recognise skin or hair problems and diseases.
- Built-in polariser filter (some models only) minimises the glaring effect of the scalp/hair
- High speed USB 2.0 connection
- Powered through USB connection: Do not need batteries
- Polariser models: Built-in 8 LEDs can be enabled or disabled as desired; an external light source is unnecessary
- Robust composite/plastic or metal housing
: Medical Device Directive 93/42/CEE modified 2007/47/EC.
: Supplied with DinoCapture (Windows) or DinoXcope software (MacOS) and carry pouch.
Frequently Bought Together