Chemical storage cabinet
Supplier: KOETTERMANN
Certificates
About this item
These cabinets are designed for the storage of non-combustible hazardous materials.
- Epoxy powder coated steel
- Lockable doors
- Optional integrated fan
- Continuous extraction inside the cabinet
- Cabinets without integrated fan should be connected to an extraction system
Maximum loading capacity: 30 kg per drawer.
Frequently Bought Together