Order Entry
Italy
ContactUsLinkComponent
Phosphatase assay kit
Phosphatase assay kit
Catalog # 786-453
Supplier:  G-Biosciences
CAS Number:  
undefined
Phosphatase assay kit
Catalog # 786-453
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Phosphatase assay kit
  • Environmentally Preferable:
  • Tests per kit:
    1000
Specifications
About this item
The Phosphatase Assay Kit is designed to measure the activity of phosphatases in biological samples and to screen for agonists and inhibitors of phosphatases. The Phosphatase Assay Kit uses para-nitrophenyl phosphate (pNPP), a chromogenic substrate for most phosphatases, including alkaline phosphatases, acid phosphatases, protein tyrosine phosphatases and serine/threonine phosphatases.
  • Colorimetric, pNPP based assay
  • Measure phosphatase activity in biological samples
  • Screen for phosphatase agonists and inhibitors
  • For the quantification of phosphatase activity
  • To screen for agonists and inhibitors of phosphatases