HPLC columns, SUPELCOGEL CA, Supelco®
Supplier: Merck
Certificates
About this item
SUPELCOGEL is a resin-based ion exclusion HPLC column which contains sulfonated divinylbenzene resins in six cationic forms. it is used for the unique selectivity for analyses of saccharides or organic acids. The SUPELCOGEL Ca columns are used to separate monosaccharides and sugar alcohols and also for separation of sugars in high fructose corn syrup.
Frequently Bought Together