Genomic DNA isolation from tissues, OmniPrep™ for Tissue kit
Catalog # 786-395
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1 KIT
- Application:
- Method / Format:PrecipitationOrganic extraction
- Prep size:Scalable
- Refine target molecule:Total DNA
- Sample type:BloodBiofluids (serum, plasma, saliva)Cultured cellsFFPE tissuesHuman/animal cells and tissues
- Target molecule:DNA
- Description:OmniPrep™ for Tissue
- Environmentally Preferable:
- Description:OmniPrep™ for Tissue
- Yield:0.5 - 10 μg/mg tissue, 25 - 50 μg/ml gram negative bacteria culture, 0.1 - 40 μg/ml body fluid
Specifications
About this item
OmniPrep™ for Tissue Kit is to specifically isolate high quality genomic DNA from tissue samples, including fresh, frozen, fixed or paraffin-embedded tissue. The kit isolates DNA from bodily fluids, including plasma, serum, amniotic fluid, semen and CSF. Also isolates high quality DNA from cultured cells, non-mammalian nucleated blood and gram-negative bacteria.
- High Yield, ~100 kb genomic DNA
- A₂₆₀/A₂₈₀ between 1,8 and 2,0
- Simple two tube method, 20 to 40 minutes
- No toxic chemicals, no phenol, no hazardous waste