Face shields, medical, 701/701.02
Supplier: UNIVET
Certificates
About this item
These acetate face shields are ideal for use in medical environments.
- Thermoformed anti-fog acetate faceshield
- Can be worn over safety spectacles
- 701 with soft foam at the front of the headband and 90° flip-up
- 701.02 with adjustable elastic band
Weight: 90 g (701) or 95 g (701.02)
Certifications: EN 166
Frequently Bought Together