Order Entry
Italy
ContactUsLinkComponent
VWR®, Hand Tally Counter
VWR®, Hand Tally Counter
  710-0935
undefined
VWR®, Hand Tally Counter
  710-0935

 

  • Range:
    0 - 9999
  • Weight:
    85 g
  • Description:
    Hand tally counter
  • W×D×H:
    30×45×45 mm
  • Environmentally Preferable:
  • Pk:
    1

 

 

Metal construction is designed for reliability, long life and hard use. Unique engineering provides a smooth tactile feel. Numbers instantly and automatically return to zero with a twist of the reset knob.

  • Compact unit for carrying in the pocket
  • Ease in viewing, large white numerals on a black background

Hundreds of laboratory applications provide opportunities to count blood samples, take surveys, inventory items, total drops of liquid, receive lab samples, and tally biomedical events. A protective window seals out dust, dirt and humidity. The rugged, heavy-duty, chrome-finished case has a hinged finger loop for a secure grip. Finger pressure on the button produces an audible click that signals the count. This feature ensures a registered count without looking at the counter.