Order Entry
Italy
ContactUsLinkComponent
(Tetrahydropyran-4-yl)methyl tosylate 98%
(Tetrahydropyran-4-yl)methyl tosylate 98%
Catalog # APOSOR11456-1G
Supplier:  Apollo Scientific
CAS Number:  
(Tetrahydropyran-4-yl)methyl tosylate 98%
Catalog # APOSOR11456-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: (Tetrahydro-2H-pyran-4-yl)methyl 4-methylbenzenesulphonate
 :
 :270.3500 g/mol
CAS Number:101691-65-0
Formula:C₁₃H₁₈O₄S
MDL Number:MFCD09475806