Sodium dodecyl sulphate (SDS), Pierce™
: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
:151-21-3
:
:CH₃(CH₂)₁₁OSO₃Na
:MFCD00036175
:1.1 g/cm³ (20 °C)
:288.38 g/mol
:204…207 °C
:>100 °C
:4.1 II
:14,08636
:205-788-1
:1325
:GHS02
:GHS06
:Ambient
- Environmentally Preferable:
Frequently Bought Together