Order Entry
Italy
ContactUsLinkComponent
Neutralising agent for the automated cleaning of glassware, neodisher® N
Neutralising agent for the automated cleaning of glassware, neodisher® N
  148-3192
Neutralising agent for the automated cleaning of glassware, neodisher® N
  148-3192
  • Description:
    neodisher® N
  • Seq:
    01
  • Environmentally Preferable:
  • Packed:
    5 litre plastic container
  • Pk:
    5 L
neodisher® N.
  • Acidic liquid concentrate for neutralisation of alkaline residues in the main automated cleaning cycle or for acidic pre-cleaning of laboratory glassware
  • Components: >30% phosphoric acid
  • Dosage: 1 to 2 ml/L for neutralisation; 3 to 4 ml/L for acidic pre-cleaning
Components: > 30% phosphoric acid
Dosage: 1 - 2 ml/l for neutralisation; 3 - 4 ml/l for acidic precleaning
Note: The materials are corrosive or irritant and care must be taken with the choice of materials for cleaning aluminium and titanium items. Please read the Material Safety Data sheet before use.
Acidic liquid concentrate for neutralisation of alakaline residues in the main automted cleaning cyle or for acidic precleaning of laboratory glassware. Free of phosphates and surfactants
Components: Organic acids
 : The materials are corrosive or irritant and care must be taken with the choice of materials for cleaning aluminium and titanium items. Please read the Material Safety Data sheet before use.