Rotors for Sorvall™ LYNX centrifuges
: SORVALL
BIOFlex™ HC Swinging-Bucket Rotor Process more than 35 tube styles from microtubes to 1L bottles, plus conical tubes, microplates, tissue culture flasks and blood bags with the Thermo Scientific™ BIOFlex™ HC Swinging-Bucket Rotor. Innovations such as Auto-Lock rotor exchange, Auto-ID instant rotor identification and speed handles on each rotor lid shorten run set-up time while providing peace of mind that the rotor is secure.
- Secure, push-button rotor exchange in less than three seconds
- Improved safety and convenience
Frequently Bought Together