Biological safety cabinets, class III, MSC III series
These biological safety cabinets have been especially designed for the handling of microbiological agents, offering maximum protection to the product, the environment and the user.
- Microprocessor air flow control
- LCD display with lighting unit constructed outside the working area
- Lexan window
- Pre-filter construction at the inlet HEPA and the exhaust HEPA
Ergonomically designed, totally sealed which keep the user completely separated from the product by a physical barrier when total containment and user protection is required.
: EN 12469.
- Catalog No:
- CLEANH336001
- CLEANH334001
- Model:
- MSC III-1800
- MSC III-1200
- Exhaust air volume:
- 0,83 m³/h
- 0,55 m³/h
- Filters:
- Inlet HEPA-filter U15 and Exhaust HEPA-filter H14 with an efficiency of 99,995% (MPPS/EN 1822)
- Inlet HEPA-filter U15 and Exhaust HEPA-filter H14 with an efficiency of 99,995% (MPPS/EN 1822)
- Lighting system:
- 800 lux
- 800 lux
- Ext. W×D×H:
- 2357×791×1240 mm
- 1757×791×1240 mm
- Int. W×D×H:
- 1794×614×870 mm
- 1194×614×870 mm
Frequently Bought Together