Filter kits, 96-well, Captiva
Supplier: VARIAN
Certificates
About this item
Faster than centrifugation and easily automated, Captiva's dual depth filtration media provide complete removal of precipitated proteins and outstanding resistance to sample clogging. The plates are also excellent for sample storage. All Captiva components are ultra clean, and rigorously tested to ensure against non specific binding.
Delivery: Each starter kit includes 5× Captiva filter plates, 10× Duo Seal 96 96-well plate seals, 5× Captiva 96-deep well 1 ml collection plates, and 5× Captiva collection plate pierceable covers. Each replacement filter kit includes 5× Captiva filter plates, 10× Duo Seal 96 96-well plate seals, 5× Captiva 96-deep well 1 ml collection plates, and 5× Captiva collection plate pierceable covers.
Frequently Bought Together