Density standard, water
Catalog # 614-1802
Supplier: Mettler - Toledo
CAS Number:
Specifications
- Pk:10
- Description:Density standard, 0,998 g/cm³, certified at 20 °C, uncertainty (typically) 0,001 g/cm³, 6 ml sachets
- Seq:10
- Environmentally Preferable: