Order Entry
Italy
ContactUsLinkComponent
 
Water DNase-, RNase-free
Catalog # J60610.XC
CAS Number:  
Water DNase-, RNase-free
Catalog # J60610.XC
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    3,8 L
  • Seq:
    5138
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Hydrogen oxide
 :
 :18.02 g/mol
Boiling Pt:100 °C
Melting Pt:0
Density:1.000
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332
Merck Index:14,10039