3,5-Dibenzyloxybenzyl bromide ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:24131-32-6
:
Formula:C₂₁H₁₉BrO₂
MDL Number:MFCD02093444
Storage Temperature:Refrigerator
UN:1759
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:85.0 to 89.0 deg-C
Specification Test Results
Frequently Bought Together