Laminated steel padlocks
Supplier: Brady
Certificates
About this item
These robust padlocks withstand severe physical abuse and come with a hardened steel shackle making them extremely difficult to cut. They are ideal for use outdoors and in high temperature environments (max. 580 °C).
- Reinforced laminated steel, with steel shackle
- Body plates are individually coated for superior rust protection
- 5-pin cylinder offers more unique key cuts and better tamper resistance than conventional 4-pin locks
- Coloured bumpers can be used for identification
- Key will not be released until the padlock shackle is closed
- Each padlock is keyed differently and is supplied with one key
Shackle diameter: 6,5 mm
Frequently Bought Together