1-Bromopropene cis- and trans-mixture ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 1-Bromo-1-propene, Propenyl bromide
CAS Number:590-14-7
:
Formula:C₃H₅Br
MDL Number:MFCD00000186
Boiling Pt:58
Molecular Weight:120.98 g/mol
Melting Pt:-76.5
Flash Pt:4°C
Storage Temperature:Refrigerator
EINECS:209-671-6
UN:1993
ADR:3 II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0% (total of isomer)
Specification Test Results
Frequently Bought Together